Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153529.2 | NP_705757.1 | 3 | 1252 | ||
GenBank mRNA | AK003046.1 | - | 3 | 1077 | ||
AK161859.1 | - | 3 | 1220 | |||
BC035531.1 | - | 3 | 1240 | 5367281 | ||
CA321643.1 | - | 1 | 645 |