Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam149a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153535.2 | NP_705763.1 | 12 | 2698 | ||
GenBank mRNA | AK034229.1 | - | 12 | 1925 | ||
AK038552.1 | - | 12 | 1922 | |||
AK140440.1 | - | 12 | 2698 | |||
AK141607.1 | - | 12 | 2641 | |||
AK181302.1 | - | 3 | 364 | |||
AK196190.1 | - | 1 | 97 | |||
AK220518.1 | - | 12 | 1943 | |||
BC035537.1 | - | 12 | 2589 | 5369152 |