Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT2B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001075.5 | NP_001066.1 | 6 | 1599 | ||
NM_001144767.2 | NP_001138239.1 | 6 | 1347 | |||
NM_001290091.1 | NP_001277020.1 | 6 | 1034 | |||
XM_017008585.1 | XP_016864074.1 | 6 | 1602 | |||
GenBank mRNA | AK222839.1 | - | 6 | 1597 | ||
AK222872.1 | - | 6 | 1601 | |||
AK292738.1 | - | 6 | 1605 | |||
AK298432.1 | - | 6 | 1346 | |||
BC113649.1 | - | 6 | 1600 | 8322701 | ||
CD013994.1 | - | 6 | 1008 |