Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phldb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153537.4 | NP_705765.3 | 22 | 4588 | ||
GenBank mRNA | AK052352.1 | - | 11 | 1948 | ||
AK122336.1 | - | 21 | 4540 | |||
BC025451.1 | - | 1 | 218 | |||
BC025856.1 | - | 4 | 786 | |||
BC053931.1 | - | 17 | 2717 | |||
BC058712.1 | - | 22 | 4584 | 6842596 | ||
BC069853.1 | - | 3 | 715 |