Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153541.3 | NP_705769.2 | 4 | 2031 | ||
GenBank mRNA | AK132152.1 | - | 4 | 2069 | ||
AK140914.1 | - | 4 | 2020 | |||
AK163930.1 | - | 4 | 2032 | |||
BC023839.1 | - | 4 | 2025 | 5344921 |