Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dennd1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153551.1 | NP_705779.1 | 19 | 1492 | ||
GenBank mRNA | BC035208.1 | - | 19 | 1492 | 3671384 |