Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Qsox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK042036.1 | - | 2, 3 | 324 | ||
AK140324.1 | - | 5, 6 | 702 | |||
AK157992.1 | - | 5, 6 | 695 | |||
AK165642.1 | - | 5, 6 | 704 | |||
BC030934.1 | - | 5, 6 | 736 | 4920915 |