Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam102a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153560.4 | NP_705788.1 | 4 | 1106 | ||
GenBank mRNA | AK049032.1 | - | 4 | 1110 | ||
AK131145.1 | - | 3 | 2824 | |||
AK157331.1 | - | 4 | 736 | |||
AK157490.1 | - | 5 | 528 | |||
AK165751.1 | - | 4 | 736 | |||
AK170536.1 | - | 4 | 1134 | |||
AK170847.1 | - | 4 | 727 | |||
AK170951.1 | - | 4 | 733 | |||
AK172536.1 | - | 4 | 733 | |||
AK183131.1 | - | 4 | 258 | |||
AK205749.1 | - | 4 | 259 | |||
BC023752.1 | - | 4 | 551 | 5354455 | ||
BC031157.1 | - | 4 | 369 | 4976773 |