Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gbp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153564.2 | NP_705792.2 | 2 | 149 | ||
GenBank mRNA | AF422243.1 | - | 2 | 131 | ||
AF487898.1 | - | 2 | 141 | |||
AK030414.1 | - | 2 | 149 | |||
AK087083.1 | - | 237 | ||||
BC058555.1 | - | 2 | 183 | 5293575 |