Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Noc4l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153570.2 | NP_705798.2 | 2 | 183 | ||
GenBank mRNA | AK052601.1 | - | 2 | 183 | ||
AK088355.1 | - | 2 | 184 | |||
AK090024.1 | - | 2 | 196 | |||
AK146006.1 | - | 2 | 179 | |||
AK172310.1 | - | 2 | 162 | |||
AK184847.1 | - | 1 | 142 | |||
BC024616.1 | - | 2 | 173 | 4218310 |