Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnot10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153585.5 | NP_705813.2 | 20 | 2887 | ||
GenBank mRNA | AK029103.1 | - | 20 | 2762 | ||
CX222096.1 | - | 2 | 476 |