Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153783.4 | NP_722478.2 | 2 | 253 | ||
GenBank mRNA | AF226656.2 | - | 2 | 272 | ||
AK030664.1 | - | 2 | 254 | |||
AM049402.1 | - | 2 | 245 | |||
BC082783.1 | - | 2 | 252 | 6475809 |