Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166653.1 | NP_001160125.1 | 2 | 295 | ||
NM_001166654.1 | NP_001160126.1 | 2 | 385 | |||
NM_001166655.1 | NP_001160127.1 | 2 | 388 | |||
NM_001166656.1 | NP_001160128.1 | 2 | 295 | |||
NM_001166657.1 | NP_001160129.1 | 2 | 385 | |||
NM_153785.4 | NP_722480.3 | 2 | 385 | |||
GenBank mRNA | AK045036.1 | - | 1 | 744 | ||
AK045356.1 | - | 2 | 285 | |||
AK046394.1 | - | 2 | 386 | |||
AK050990.1 | - | 2 | 385 | |||
AK078804.1 | - | 2 | 277 | |||
AK090045.1 | - | 2 | 171 | |||
AK186403.1 | - | 2 | 118 | |||
AK210497.1 | - | 2 | 179 | |||
BC028871.1 | - | 2 | 257 | 4241862 |