Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000930.4 | NP_000921.1 | 14 | 2511 | ||
NM_001319189.1 | NP_001306118.1 | 12 | 2244 | |||
NM_033011.3 | NP_127509.1 | 13 | 2373 | |||
GenBank mRNA | A03776.1 | - | 13 | 2288 | ||
BC002795.2 | - | 13 | 2199 | 3618149 | ||
BC007231.1 | - | 14 | 2335 | 3161005 | ||
BC013968.2 | - | 14 | 2337 | 3163742 | ||
BC018636.2 | - | 14 | 2337 | 3637273 | ||
BC095403.1 | - | 14 | 2352 | 30528441 | ||
D01096.1 | - | 12 | 2168 | |||
M15518.1 | - | 14 | 2328 | |||
M18182.1 | - | 13 | 2267 | |||
X02901.1 | - | 13 | 2268 | |||
X07393.1 | - | 14 | 2307 | |||
X13097.1 | - | 11 | 1847 |