Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ank3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009670.4 | NP_033800.2 | 4 | 912 | ||
NM_146005.3 | NP_666117.2 | 27 | 3383 | |||
NM_170687.3 | NP_733788.2 | 4 | 912 | |||
NM_170688.2 | NP_733789.1 | 27 | 3383 | |||
NM_170689.2 | NP_733790.2 | 26 | 3329 | |||
NM_170690.2 | NP_733791.2 | 26 | 3320 | |||
NM_170728.2 | NP_733924.2 | 25 | 3266 | |||
NM_170729.2 | NP_733925.2 | 25 | 3266 | |||
NM_170730.2 | NP_733926.2 | 2 | 602 | |||
GenBank mRNA | AF212924.1 | - | 2 | 102 | ||
AK035681.1 | - | 4 | 640 | |||
AK141720.1 | - | 26 | 3205 | |||
AK157735.1 | - | 4 | 627 | |||
AK162007.1 | - | 4 | 570 | |||
BC021657.1 | - | 25 | 3136 | 4188590 | ||
BC026152.1 | - | 4 | 513 | |||
L40631.1 | - | 4 | 915 | |||
L40632.1 | - | 27 | 2925 | |||
U89274.1 | - | 6 | 529 | |||
U89275.1 | - | 6 | 529 |