Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170758.3 | NP_739564.1 | 2 | 438 | ||
GenBank mRNA | AB065156.1 | - | 2 | 157 | ||
AB091765.1 | - | 2 | 176 | |||
AB091766.1 | - | 2 | 160 | |||
AK045869.1 | - | 2 | 438 | |||
AK053160.1 | - | 2 | 126 | |||
AY457054.1 | - | 2 | 114 | |||
BC139296.1 | - | 2 | 374 | 8862318 | ||
BC139297.1 | - | 2 | 374 | 8862319 |