Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153578.2 | NP_705806.1 | 1, 2 | 173 | ||
GenBank mRNA | AK046619.1 | - | 1, 2 | 188 | ||
AK166325.1 | - | 151 | ||||
AY098645.1 | - | 1, 2 | 173 | |||
BC055828.1 | - | 1, 2 | 173 | 4192279 |