Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6ka5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153587.2 | NP_705815.1 | 3 | 408 | ||
GenBank mRNA | AK032316.1 | - | 3 | 501 | ||
AK046319.1 | - | 3 | 185 | |||
AK135786.1 | - | 316 | ||||
AY341873.1 | - | 3 | 397 | |||
AY341881.1 | - | 3 | 397 | |||
BC035298.1 | - | 3 | 408 | 4021865 |