Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153592.2 | NP_705820.1 | 6 | 470 | ||
GenBank mRNA | AK030557.1 | - | 6 | 414 | ||
BC036333.1 | - | 6 | 435 | 3590904 | ||
BC038374.1 | - | 6 | 410 | 5355446 |