Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcmtd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291211.1 | NP_001278140.1 | 7 | 1233 | ||
NM_153594.3 | NP_705822.1 | 7 | 1229 | |||
GenBank mRNA | AB182640.1 | - | 7 | 1174 | ||
AK031335.1 | - | 7 | 1184 | |||
AK077319.1 | - | 2 | 273 | |||
AK079927.1 | - | 4 | 505 | |||
AK088800.1 | - | 7 | 1168 | |||
AK146080.1 | - | 7 | 1176 | |||
BC024721.1 | - | 1 | 373 | |||
BC024791.1 | - | 2 | 323 | |||
BC031360.1 | - | 6 | 1046 | 5359242 | ||
BC040385.1 | - | 7 | 1180 | 3499499 |