Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000942.4 | NP_000933.1 | 3 | 430 | ||
GenBank mRNA | AK291149.1 | - | 3 | 294 | ||
BC001125.2 | - | 3 | 279 | 2966791 | ||
BC008848.2 | - | 3 | 401 | 3502055 | ||
BC020800.1 | - | 3 | 286 | 4248828 | ||
BC032138.2 | - | 3 | 284 | 4896656 | ||
BQ931949.1 | - | 3 | 497 | |||
CR456829.1 | - | 3 | 261 | |||
EU794651.1 | - | 3 | 430 | |||
M60457.1 | - | 3 | 235 | |||
M60857.1 | - | 3 | 249 | |||
M63573.1 | - | 3 | 282 | |||
U95646.1 | - | 706 |