Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bclaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025392.1 | NP_001020563.1 | 4 | 392 | ||
NM_001025393.1 | NP_001020564.1 | 4 | 392 | |||
NM_153787.2 | NP_722482.1 | 4 | 386 | |||
GenBank mRNA | AK011802.1 | - | 4 | 332 | ||
AK088741.1 | - | 4 | 327 | |||
AK141834.1 | - | 4 | 339 | |||
AK149724.1 | - | 4 | 345 | |||
AK150017.1 | - | 4 | 345 | |||
AK161692.1 | - | 4 | 390 | |||
AK162896.1 | - | 4 | 350 | |||
BC028963.1 | - | 4 | 324 | |||
BC029682.1 | - | 4 | 323 | |||
BC034300.1 | - | 4 | 318 | 4507876 | ||
BC086624.1 | - | 4 | 377 | |||
CK637794.1 | - | 1 | 48 |