Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153795.2 | NP_722490.1 | 4 | 694 | ||
GenBank mRNA | AK090332.1 | - | 4 | 591 | ||
AK142912.1 | - | 4 | 695 | |||
AK149737.1 | - | 4 | 625 | |||
AK158357.1 | - | 4 | 590 | |||
AK169682.1 | - | 4 | 605 | |||
BC025119.1 | - | 4 | 626 | 4190760 | ||
BC032204.1 | - | 4 | 670 | 5342463 | ||
BC066803.1 | - | 4 | 655 | 30043017 |