Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153800.4 | NP_722495.3 | 9 | 1123 | ||
GenBank mRNA | AK046097.1 | - | 9 | 1125 | ||
AK052541.1 | - | 6 | 758 | |||
AK078218.1 | - | 8 | 1012 | |||
AY541447.1 | - | 5 | 956 | |||
BC038272.1 | - | 4 | 621 | 4237331 |