Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153801.3 | NP_722496.2 | 1, 2 | 431 | ||
GenBank mRNA | AF548365.1 | - | 1, 2 | 432 | ||
AK052276.1 | - | 1, 2 | 270 | |||
AK052284.1 | - | 1, 2 | 434 | |||
AK052302.1 | - | 1, 2 | 136 | |||
AK052378.1 | - | 1, 2 | 194 | |||
BC107366.1 | - | 1, 2 | 344 | 40056219 | ||
BC107367.1 | - | 1, 2 | 344 | 40056223 |