Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glb1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153803.1 | NP_722498.1 | 2, 3 | 186 | ||
GenBank mRNA | BC038479.1 | - | 2, 3 | 186 | 4503030 |