Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000948.5 | NP_000939.1 | 5 | 1315 | ||
NM_001163558.2 | NP_001157030.1 | 6 | 987 | |||
XM_011514753.2 | XP_011513055.1 | 6 | 1034 | |||
XM_011514754.2 | XP_011513056.1 | 7 | 1047 | |||
GenBank mRNA | BC015850.1 | - | 5 | 1318 | 4657724 | |
BC088370.1 | - | 5 | 845 | 30393428 | ||
CD106243.1 | - | 2 | 325 | |||
D00411.1 | - | 4 | 768 | |||
GQ305133.1 | - | 6 | 987 | |||
M29386.1 | - | 4 | 768 | |||
V00566.1 | - | 5 | 800 | |||
X54393.1 | - | 6 | 937 |