Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRLR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000949.6 | NP_000940.1 | 9 | 1267 | ||
NM_001204314.2 | NP_001191243.1 | 6 | 642 | |||
NM_001204315.1 | NP_001191244.1 | 8 | 945 | |||
NM_001204316.1 | NP_001191245.1 | 8 | 945 | |||
NM_001204317.1 | NP_001191246.1 | 8 | 945 | |||
NR_037910.1 | 7 | 812 | ||||
XM_006714484.2 | XP_006714547.1 | 9 | 1085 | |||
XM_011514068.2 | XP_011512370.1 | 9 | 1100 | |||
XM_011514069.2 | XP_011512371.1 | 9 | 1039 | |||
XM_017009645.1 | XP_016865134.1 | 8 | 1033 | |||
XM_017009646.1 | XP_016865135.1 | 7 | 883 | |||
GenBank mRNA | AF166329.1 | - | 7 | 840 | ||
AF349939.1 | - | 5 | 537 | |||
AF416618.1 | - | 7 | 857 | |||
AF416619.1 | - | 7 | 857 | |||
AF493069.1 | - | 6 | 724 | |||
AF512796.1 | - | 6 | 531 | |||
AK313270.1 | - | 9 | 1155 | |||
BC059392.1 | - | 9 | 1153 | 30343995 | ||
GU133399.1 | - | 7 | 857 | |||
M31661.1 | - | 9 | 1124 |