Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nudt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291044.1 | NP_001277973.1 | 5 | 857 | ||
NM_153561.3 | NP_705789.1 | 5 | 854 | |||
GenBank mRNA | AF453427.1 | - | 5 | 966 | ||
AF453428.1 | - | 5 | 843 | |||
AK145299.1 | - | 5 | 849 | |||
BC027267.1 | - | 5 | 848 | 3500531 | ||
BC146000.1 | - | 5 | 851 | 40131303 |