Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153573.1 | NP_705801.1 | 3, 4 | 570 | ||
GenBank mRNA | AK030620.1 | - | 3, 4 | 621 | ||
AK081692.1 | - | 3, 4 | 597 | |||
AK163384.1 | - | 716 | ||||
BC024846.1 | - | 997 | ||||
BC029109.1 | - | 3, 4 | 570 | 3985764 | ||
BC034313.1 | - | 4, 5 | 685 | 5365046 |