Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sv2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109753.1 | NP_001103223.1 | 11 | 2168 | ||
NM_153579.4 | NP_705807.2 | 11 | 2375 | |||
GenBank mRNA | AK029736.1 | - | 12 | 2200 | ||
AK043830.1 | - | 11 | 2095 | |||
AK122359.1 | - | 10 | 1320 | |||
AK139478.1 | - | 11 | 2070 | |||
AK140104.1 | - | 11 | 2088 | |||
AK147261.1 | - | 11 | 2162 | |||
AK159028.1 | - | 11 | 2073 | |||
BC034651.1 | - | 1 | 104 | |||
BC060224.1 | - | 10 | 1759 | 30356990 | ||
BC079568.1 | - | 11 | 2375 |