Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Erlin2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153592.2 | NP_705820.1 | 12 | 2186 | ||
GenBank mRNA | AK030557.1 | - | 12 | 2130 | ||
AK160391.1 | - | 1 | 331 | |||
AK160885.1 | - | 1 | 1061 | |||
BC024060.1 | - | 4 | 1466 | |||
BC038374.1 | - | 12 | 2126 | 5355446 |