Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpt1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252470.1 | NP_001239399.1 | 15, 16 | 2130 | ||
NM_153679.2 | NP_710146.1 | 15, 16 | 2132 | |||
GenBank mRNA | AF320000.1 | - | 15, 16 | 2055 | ||
AK035790.1 | - | 15, 16 | 2096 | |||
BC066155.1 | - | 15, 16 | 2054 | 30089333 | ||
BC083317.1 | - | 15, 16 | 2013 |