Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153680.2 | NP_710147.1 | 15 | 1556 | ||
GenBank mRNA | AK047265.1 | - | 11 | 1950 | ||
AK077650.1 | - | 15 | 1558 | |||
AK077780.1 | - | 15 | 1558 | |||
AK156299.1 | - | 15 | 1607 | |||
BC023732.1 | - | 15 | 1572 | 5352355 | ||
BC023925.1 | - | 10 | 1019 | |||
BC026571.1 | - | 15 | 1556 | 4986268 | ||
BC029675.1 | - | 15 | 1541 |