Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atoh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153778.3 | NP_722473.1 | 3 | 2075 | ||
GenBank mRNA | AB046527.1 | - | 3 | 2078 | ||
AB046528.1 | - | 3 | 1774 | |||
AB046529.1 | - | 3 | 1036 | |||
AB046530.1 | - | 4 | 1188 | |||
AK016909.1 | - | 3 | 1026 | |||
AY349615.1 | - | 5 | 1751 | |||
BC023684.1 | - | 3 | 1743 | 5343472 |