Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paox | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153783.4 | NP_722478.2 | 3 | 742 | ||
GenBank mRNA | AF226656.2 | - | 3 | 761 | ||
AK010469.1 | - | 2 | 391 | |||
AK030664.1 | - | 3 | 743 | |||
AM049402.1 | - | 3 | 734 | |||
BC082783.1 | - | 3 | 741 | 6475809 |