Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fermt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153795.2 | NP_722490.1 | 14 | 1905 | ||
GenBank mRNA | AK090332.1 | - | 14 | 1802 | ||
AK142912.1 | - | 14 | 1906 | |||
AK149737.1 | - | 13 | 2328 | |||
AK158357.1 | - | 14 | 1802 | |||
AK169682.1 | - | 14 | 1816 | |||
BC025119.1 | - | 14 | 1837 | 4190760 | ||
BC032204.1 | - | 14 | 1881 | 5342463 | ||
BC066803.1 | - | 13 | 2187 | 30043017 |