Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tecrl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153801.3 | NP_722496.2 | 6, 7 | 854 | ||
GenBank mRNA | AF548365.1 | - | 6, 7 | 855 | ||
AK052276.1 | - | 6, 7 | 693 | |||
AK052284.1 | - | 6, 7 | 857 | |||
AK052302.1 | - | 6, 7 | 559 | |||
AK052378.1 | - | 6, 7 | 617 | |||
BC107366.1 | - | 6, 7 | 767 | 40056219 | ||
BC107367.1 | - | 6, 7 | 767 | 40056223 |