Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252684.1 | NP_001239613.1 | 3 | 448 | ||
NM_001252685.1 | NP_001239614.1 | 3 | 277 | |||
NM_153808.2 | NP_722503.1 | 3 | 448 | |||
GenBank mRNA | AJ534941.1 | - | 3 | 369 | ||
AK143818.1 | - | 3 | 425 | |||
AK161254.1 | - | 3 | 278 | |||
BC038345.1 | - | 3 | 428 | 3596405 |