Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd300a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170758.3 | NP_739564.1 | 7 | 2136 | ||
GenBank mRNA | AK045869.1 | - | 7 | 2136 | ||
BC139296.1 | - | 7 | 2072 | 8862318 | ||
BC139297.1 | - | 7 | 2072 | 8862319 |