Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168684.1 | NP_001162155.1 | 2 | 290 | ||
NM_172051.3 | NP_742048.2 | 2 | 367 | |||
NM_177026.1 | NP_796000.1 | 2 | 309 | |||
GenBank mRNA | AK079611.1 | - | 3 | 227 | ||
AK083129.1 | - | 2 | 270 | |||
AK132545.1 | - | 2 | 345 | |||
BB619673.1 | - | 2 | 368 | |||
BC026867.1 | - | 2 | 290 | 3660220 | ||
BC057202.1 | - | 2 | 153 | 5327878 | ||
BC095951.1 | - | 2 | 301 | 5034018 | ||
BY145366.1 | - | 2 | 310 |