Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172054.4 | NP_742051.1 | 2 | 299 | ||
GenBank mRNA | AK002893.1 | - | 2 | 304 | ||
AK006170.1 | - | 2 | 354 | |||
AK010709.1 | - | 2 | 298 | |||
AK011424.1 | - | 2 | 243 | |||
AK016756.1 | - | 2 | 286 | |||
AK028525.1 | - | 2 | 296 | |||
AK153857.1 | - | 2 | 300 | |||
AK167045.1 | - | 2 | 263 | |||
BC022947.1 | - | 2 | 280 | 3584012 | ||
BC083077.1 | - | 2 | 296 | 6466275 |