Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hinfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172162.3 | NP_751894.2 | 2 | 163 | ||
GenBank mRNA | AK049348.1 | - | 2 | 127 | ||
AK049633.1 | - | 2 | 324 | |||
AK167360.1 | - | 3 | 280 | |||
BC029091.1 | - | 1 | 146 | 2651006 | ||
BY196765.1 | - | 2 | 163 | |||
CT010239.1 | - | 1 | 98 |