Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284346.1 | NP_001271275.1 | 1, 2 | 291 | ||
NM_172254.4 | NP_758458.1 | 1, 2 | 291 | |||
GenBank mRNA | AK011448.1 | - | 1, 2 | 209 | ||
AK048097.1 | - | 1, 2 | 181 | |||
BC029910.1 | - | 1, 2 | 182 | 4920779 | ||
BC039954.1 | - | 1, 2 | 202 | 4193739 |