Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172086.2 | NP_742083.1 | 4 | 442 | ||
GenBank mRNA | AK002353.1 | - | 4 | 443 | ||
AK011017.1 | - | 4 | 443 | |||
AK012525.1 | - | 4 | 443 | |||
AK028204.1 | - | 4 | 443 | |||
AK146768.1 | - | 4 | 443 | |||
AK148453.1 | - | 4 | 443 | |||
AK150705.1 | - | 4 | 443 | |||
AK168282.1 | - | 4 | 446 | |||
BC030176.1 | - | 441 | ||||
BC046339.1 | - | 4 | 411 | 6478080 | ||
BC086909.1 | - | 4 | 446 | 30122780 |
Gene Symbol: Gm6251 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001101561.2 | NP_001095031.2 | 1 | 436 | ||
GenBank mRNA | CN721608.1 | - | 1 | 436 |