Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000989.3 | NP_000980.1 | 4, 5 | 421 | ||
GenBank mRNA | AK312102.1 | - | 4, 5 | 374 | ||
BC032700.2 | - | 3, 4 | 349 | 5393199 | ||
BC095426.1 | - | 4, 5 | 370 | 30792764 | ||
BM785413.1 | - | 4, 5 | 421 | |||
BQ055546.1 | - | 3, 4 | 323 | |||
L05095.1 | - | 4, 5 | 360 | |||
X79238.1 | - | 3, 4 | 306 |