Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sidt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289668.1 | NP_001276597.1 | 2, 3 | 652 | ||
NM_172257.4 | NP_758461.1 | 2, 3 | 652 | |||
GenBank mRNA | AK081177.1 | - | 2, 3 | 638 | ||
AK152371.1 | - | 2, 3 | 585 | |||
AK155875.1 | - | 2, 3 | 556 | |||
AK196459.1 | - | 2, 3 | 184 | |||
BC023957.1 | - | 2, 3 | 656 | 5321190 | ||
BC051101.1 | - | 2, 3 | 380 | 5052849 | ||
CB247121.1 | - | 2, 3 | 661 |