Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL36AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001.4 | NP_000992.1 | 2 | 553 | ||
GenBank mRNA | AB063609.1 | - | 2 | 499 | ||
BC000741.2 | - | 2 | 486 | 3507009 | ||
BC003145.1 | - | 2 | 479 | 2957923 | ||
BC070207.1 | - | 2 | 488 | 4271403 | ||
BC107716.1 | - | 2 | 513 | 6727975 |