Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plcg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172285.1 | NP_758489.1 | 2, 3 | 390 | ||
GenBank mRNA | AK142173.1 | - | 2, 3 | 393 | ||
AK151039.1 | - | 2, 3 | 376 | |||
AK170430.1 | - | 2, 3 | 392 | |||
BC023877.1 | - | 2, 3 | 390 | 5354925 |