Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172301.3 | NP_758505.2 | 3 | 312 | ||
GenBank mRNA | AK136259.1 | - | 3 | 326 | ||
AK143597.1 | - | 3 | 268 | |||
AK163259.1 | - | 3 | 325 | |||
AK166686.1 | - | 3 | 326 | |||
AK168441.1 | - | 3 | 326 | |||
AK168502.1 | - | 3 | 326 | |||
AK168993.1 | - | 3 | 323 | |||
BC011478.1 | - | 3 | 312 | 4014448 | ||
BC080202.1 | - | 3 | 282 | 3603637 | ||
BC085238.1 | - | 3 | 308 | 6337228 | ||
FJ931532.1 | - | 3 | 228 | |||
S43105.1 | - | 3 | 350 | |||
X58708.1 | - | 3 | 316 | |||
X64713.1 | - | 3 | 303 |